Anti-MAPT Rabbit Polyclonal Antibody

NBP1-51308
102181-384EA 466.15 USD
102181-384
Anti-MAPT Rabbit Polyclonal Antibody
Rabbit Polyclonal Tau Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR