Anti-TATA Element Modulatory Factor 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79708
102215-064EA 564.74 USD
102215-064
Anti-TATA Element Modulatory Factor 1 Rabbit Polyclonal Antibody
Antibodies
The TATA Element Modulatory Factor 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TATA Element Modulatory Factor 1. This antibody reacts with human. The TATA Element Modulatory Factor 1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TATA Element Modulatory Factor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR