Anti-TDP-43/TARDBP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Novus Biologicals
NB110-55376H
102122-594EA 614.15 USD
102122-594
Anti-TDP-43/TARDBP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The TDP-43 / TARDBP Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to TDP-43 / TARDBP. This antibody reacts with human, mouse, chicken, primate, xenopus, zebrafish. The TDP-43 / TARDBP Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: TDP-43/TARDBP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken, Primate, Xenopus, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR