Anti-TAP1 Rabbit Polyclonal Antibody

NBP1-59030
102191-776EA 466.15 USD
102191-776
Anti-TAP1 Rabbit Polyclonal Antibody
Rabbit Polyclonal TAP1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Chicken, Xenopus.

Type: Primary
Antigen: TAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR