Anti-TNKS Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB100-186B
102106-216EA 323 USD
102106-216
Anti-TNKS Rabbit Polyclonal Antibody (Biotin)
The Tankyrase 1 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to Tankyrase 1. This antibody reacts with human. The Tankyrase 1 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TNKS
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR