Anti-TANK Rabbit Polyclonal Antibody

NBP1-76716
102209-088EA 466.15 USD
102209-088
Anti-TANK Rabbit Polyclonal Antibody
The TANK Antibody from Novus Biologicals is a rabbit polyclonal antibody to TANK. This antibody reacts with human, mouse, rat. The TANK Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TANK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR