Anti-TANC2 Rabbit Polyclonal Antibody

NBP1-52634
102182-992EA 507.69 USD
102182-992
Anti-TANC2 Rabbit Polyclonal Antibody
Rabbit Polyclonal TANC2 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: TANC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR