Anti-TAF7L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80292
102216-232EA 564.74 USD
102216-232
Anti-TAF7L Rabbit Polyclonal Antibody
Antibodies
The TAF7L Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAF7L. This antibody reacts with mouse. The TAF7L Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TAF7L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR