Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAF6L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74133
The TAF6L Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAF6L. This antibody reacts with rat. The TAF6L Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TAF6L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-74133 102205-008 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TAF6L
Antigen Name TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa
Reactivity Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 6L, PCAF-associated factor 65-alpha, p300/CBP-associated factor (PCAF)-associated factor 65, PAF65AMGC4288, PCAF-associated factor 65 alpha, FLJ11136, 65kDa, 65 kD, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, PAF65-alpha
Gene ID 10629
Immunogen Synthetic peptides corresponding to the middle region of Taf6l. Immunizing peptide sequence PQLMKVALQDLQTNSKIAALLPYFVYVVSGVKSVSHDLEQLHRLLQVARS.
Purification Immunogen affinity purified
Storage Buffer PBS & 2% Sucrose.
Storage Temperature Store at -20C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping