Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAF6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-60639
The TAF6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAF6. This antibody reacts with human. The TAF6 Antibody has been validated for the following applications: Immunoprecipitation.

Type: Primary
Antigen: TAF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NB100-60639 102111-036 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TAF6
Antigen Name TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoPrecipitation Yes
Antigen Synonyms Transcription initiation factor TFIID 70 kDa subunit, TAF(II)70, TATA box binding protein (TBP)-associated factor, TAFII70TAFII-80, TAFII85, TAF2ERNA polymerase II TBP-associated factor subunit E, DKFZp781E21155, 80kDa, TAF(II)80, 70/85kD, RNA polymerase II, TAF6 RNA polymerase II, transcription initiation factor TFIID subunit 6, Transcription initiation factor TFIID 80 kDa subunit, MGC:8964, TAFII80TAFII-70, E
Gene ID 6878
Concentration 1.0 mg/ml
Immunogen The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human TBP-associated factor 6 using the numbering given in entry NP_005632.1 (GeneID 6878).
Purification Immunogen affinity purified
Storage Buffer Tris-Citrate/Phosphate (pH 7.0 - 8.0)
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty