Anti-TAF2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-21372
102160-252EA 635.51 USD
102160-252
Anti-TAF2 Rabbit Polyclonal Antibody
Antibodies
The TAF2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAF2. This antibody reacts with human. The TAF2 Antibody has been validated for the following applications: Immunoprecipitation, Western Blot (Negative).

Type: Primary
Antigen: TAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR