Anti-TAF15 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-567
102110-234EA 635.51 USD
102110-234
Anti-TAF15 Rabbit Polyclonal Antibody
Antibodies
The TAF15 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAF15. This antibody reacts with human. The TAF15 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TAF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR