Anti-TACC2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31221
102164-314EA 635.51 USD
102164-314
Anti-TACC2 Rabbit Polyclonal Antibody
Antibodies
The TACC2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TACC2. This antibody reacts with human. The TACC2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TACC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR