Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-21405
The TAB1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAB1. This antibody reacts with human, mouse. The TAB1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-21405 102160-306 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TAB1
Antigen Name TGF-Beta Activated Kinase 1
Reactivity Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoPrecipitation Yes
Antigen Synonyms Mitogen-activated protein kinase kinase kinase 7-interacting protein 1, TGF-beta-activated kinase 1-binding protein 1, MGC57664, transforming growth factor beta-activated kinase-binding protein 1, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, TGF-beta-activated kinase 1 and MAP3K7-binding protein 1, TAK1-binding protein 1, TGF-beta activated kinase 1/MAP3K7 binding protein 1, MAP3K7IP13'-Tab1
Gene ID 10454
Concentration 0.2 mg/ml
Immunogen The immunogen recognized by this antibody maps to a region between residue 435 and 485 of human TAK1-binding protein 1 using the numbering given in entry NP_006107.1 (GeneID 10454).
Purification Immunogen affinity purified
Storage Buffer TBS and 0.1% BSA
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty