Anti-TAAR5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NLS319
102253-254EA 329 USD
102253-254
Anti-TAAR5 Rabbit Polyclonal Antibody
Antibodies
The TAAR5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAAR5. This antibody reacts with human, monkey. The TAAR5 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TAAR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR