Anti-T7 Epitope Tag Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB600-372
102133-850EA 299 USD
102133-850
Anti-T7 Epitope Tag Rabbit Polyclonal Antibody
Antibodies
The T7 Epitope Tag Antibody from Novus Biologicals is a rabbit polyclonal antibody to T7 Epitope Tag. This antibody reacts with non-species specific. The T7 Epitope Tag Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: T7 Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR