Anti-T7 Epitope Tag Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB600-372B
102133-852EA 399 USD
102133-852
Anti-T7 Epitope Tag Rabbit Polyclonal Antibody (Biotin)
Antibodies
The T7 Epitope Tag Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to T7 Epitope Tag. This antibody reacts with non-species specific. The T7 Epitope Tag Antibody [Biotin] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: T7 Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR