Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-T7 Epitope Tag Rabbit Polyclonal Antibody (Agarose Immobilized)

Supplier: Novus Biologicals
NBP1-40393
The T7 Epitope Tag Antibody [Agarose Immobilized] from Novus Biologicals is a rabbit polyclonal antibody to T7 Epitope Tag. This antibody reacts with non-species specific. The T7 Epitope Tag Antibody [Agarose Immobilized] has been validated for the following applications: Immunoprecipitation.

Type: Primary
Antigen: T7 Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation: Agarose Immobilized
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Non-species specific




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NBP1-40393 102169-294 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol T7 Epitope Tag
Antigen Name T7 Epitope Tag
Reactivity Non-species specific
Conjugation Agarose Immobilized
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoPrecipitation Yes
Antigen Synonyms T7 epitope tag, MASMTGGQQMG tag, MASMTGGQQMG epitope tag
Immunogen Rabbits were immunized with T7 cleavage site (MASMTGGQQMG) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. Rabbit anti- human T7 affinity purified antibodies were coupled to peptide immobilized on solid support.
Purification Immunogen affinity purified
Storage Buffer PBS
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty