Anti-Syntenin 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31136
102164-186EA 635.51 USD
102164-186
Anti-Syntenin 1 Rabbit Polyclonal Antibody
Antibodies
The Syntenin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Syntenin 1. This antibody reacts with human, mouse, zebrafish. The Syntenin 1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Syntenin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR