Anti-Syntaxin 1A Rabbit Polyclonal Antibody

NBP1-98478
102223-272EA 466.15 USD
102223-272
Anti-Syntaxin 1A Rabbit Polyclonal Antibody
The Syntaxin 1A Antibody from Novus Biologicals is a rabbit polyclonal antibody to Syntaxin 1A. This antibody reacts with mouse. The Syntaxin 1A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Syntaxin 1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR