Anti-Synaptotagmin VI Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-11539
102249-862EA 564.62 USD
102249-862
Anti-Synaptotagmin VI Rabbit Polyclonal Antibody
Antibodies
The Synaptotagmin VI Antibody from Novus Biologicals is a rabbit polyclonal antibody to Synaptotagmin VI. This antibody reacts with human. The Synaptotagmin VI Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: Synaptotagmin VI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR