Anti-SYT Rabbit Polyclonal Antibody

NBP1-19363
102158-724EA 466.15 USD
102158-724
Anti-SYT Rabbit Polyclonal Antibody
Rabbit Polyclonal Synaptotagmin 1 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200

Type: Primary
Antigen: SYT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR