Anti-Synaptophysin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59660
102193-008EA 564.74 USD
102193-008
Anti-Synaptophysin Rabbit Polyclonal Antibody
Antibodies
The Synaptophysin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Synaptophysin. This antibody reacts with human, mouse, rat. The Synaptophysin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Synaptophysin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR