Anti-Symplekin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-74592
102116-524EA 635.51 USD
102116-524
Anti-Symplekin Rabbit Polyclonal Antibody
Antibodies
The Symplekin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Symplekin. This antibody reacts with human. The Symplekin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Symplekin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR