Anti-Swi6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB600-932
102134-742EA 777.05 USD
102134-742
Anti-Swi6 Rabbit Polyclonal Antibody
Antibodies
The Swi6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Swi6. This antibody reacts with yeast. The Swi6 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Swi6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR