|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SUV420h1 | |
Antigen Name | Suppressor of Variegation 4-20 Homolog 1 | |
Reactivity | Mouse, Human | Human, Mouse |
Conjugation | Unconjugated | |
Clonality | Polyclonal | |
Antibody Type | Primary | |
Host | Rabbit | |
Isotype | IgG | |
Western Blot | Yes | |
ImmunoChemistry | Yes | |
ImmunoFluorescence | Yes | |
Antigen Synonyms | Su(var)4-20 homolog 1, CGI85, histone-lysine N-methyltransferase SUV420H1, MGC118909, KMT5B Lysine N-methyltransferase 5B, Suppressor of variegation 4-20 homolog 1, EC 2.1.1.43, CGI-85, MGC21161, MGC703, C630029K18Rik, MGC118906, suv4-20h1, KMT5B, suppressor of variegation 4-20 homolog 1 (Drosophila) | Su(var)4-20 homolog 1, CGI85, Suppressor of variegation 4-20 homolog 1, EC 2.1.1.43, suppressor of variegation 4-20 homolog 1 (Drosophila), KMT5B, suv4-20h1, CGI-85, KMT5B Lysine N-methyltransferase 5B, MGC21161, C630029K18Rik, MGC118906, histone-lysine N-methyltransferase SUV420H1, MGC118909, MGC703 |
Gene ID | 51111 | |
Concentration | 1.16 mg/ml | |
Immunogen | A synthetic peptide made to an internal portion of the human SUV420h1 protein (between residues 450-500) [UniProt Q4FZB7] | |
Purification | Immunogen affinity purified | |
Storage Buffer | PBS and 30% Glycerol | |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |