Anti-BIRC5 Rabbit Polyclonal Antibody

NBP1-76840
102209-572EA 466.15 USD
102209-572
Anti-BIRC5 Rabbit Polyclonal Antibody
Rabbit Polyclonal Survivin Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR