|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | BIRC5 |
Antigen Name | Survivin |
Reactivity | Hamster, Feline, Rat, Mouse, Canine, Guinea Pig, Human |
Conjugation | DyLight® 550 |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoPrecipitation | Yes |
ImmunoFluorescence | Yes |
Flow Cytometry | Yes |
Antigen Synonyms | Apoptosis inhibitor survivin, IAP4, API4 survivin variant 3 alpha, BIRC5, baculoviral IAP repeat-containing 5, Apoptosis inhibitor 4 baculoviral IAP repeat-containing protein 5, EPR-1 |
Gene ID | 332 |
Immunogen | Full length recombinant human Survivin [UniProt# O15392] |
Purification | Immunogen affinity purified |
Storage Buffer | 50mM Sodium Borate |
Storage Temperature | Store at 4C in the dark. |