Anti-Survivin Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB500-236R
102132-060EA 614.15 USD
102132-060
Anti-Survivin Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The Survivin [p Thr34] Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to Survivin. This antibody reacts with human, mouse. The Survivin [p Thr34] Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Survivin
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR