Anti-SUPT5H Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-40592
102121-944EA 635.51 USD
102121-944
Anti-SUPT5H Rabbit Polyclonal Antibody
Antibodies
The SUPT5H Antibody from Novus Biologicals is a rabbit polyclonal antibody to SUPT5H. This antibody reacts with human. The SUPT5H Antibody has been validated for the following applications: Immunoprecipitation, Western Blot (Negative).

Type: Primary
Antigen: SUPT5H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR