Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SULT1C4 Rabbit Polyclonal Antibody (Unconjugated) [clone: ]

Supplier: Novus Biologicals
NBP1-52893
The Cytosolic Sulfotransferase 1C4 / SULT1C4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cytosolic Sulfotransferase 1C4 / SULT1C4. This antibody reacts with human. The Cytosolic Sulfotransferase 1C4 / SULT1C4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SULT1C4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-52893 102183-252 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SULT1C4
Antigen Name Cytosolic Sulfotransferase 1C4
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms SULT1C2cytosolic, EC 2.8.2.1, member 2, SULT1C#2, MGC34422, Sulfotransferase 1C2, cytosolic, ST1C4, member C2, 1C, sulfotransferase family, EC 2.8.2, EC 2.8.2.-, member 4, MGC149521
Gene ID 27233
Immunogen Synthetic peptides corresponding to SULT1C4(sulfotransferase family, cytosolic, 1C, member 4) The peptide sequence was selected from the middle region of SULT1C4. Peptide sequence HEHVKGWWEAKDKHRILYLFYEDMKKNPKHEIQKLAEFIGKKLDDKVLDK.
Purification Immunogen affinity purified
Storage Buffer PBS and 2% Sucrose
Storage Temperature Store at -20C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping