Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SULT1B1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54829
102184-620
Anti-SULT1B1 Rabbit Polyclonal Antibody
The Cytosolic Sulfotransferase 1B1 / SULT1B1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cytosolic Sulfotransferase 1B1 / SULT1B1. This antibody reacts with human. The Cytosolic Sulfotransferase 1B1 / SULT1B1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SULT1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty