|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SUG1 |
Reactivity | Mouse, Human, Rat |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | MSUG1 protein, Proteasome 26S subunit ATPase 5, Proteasome subunit p45, p45/SUG26S protease regulatory subunit 8, ATPase, proteasome 26S ATPase subunit 5, TBP10, macropain) 26S subunit, p45, thyroid receptor interactor 1, S8, 5, SUG126S proteasome AAA-ATPase subunit RPT6, proteasome (prosome, SUG-1, Tat-binding protein homolog 10, TRIP1Thyroid hormone receptor-interacting protein 1 |
Gene ID | 5705 |
Concentration | 1 mg/ml |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PSMC5. The exact sequence is proprietary. |
Purification | Immunogen affinity purified |
Storage Buffer | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |