Anti-SUB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-59775
102110-770EA 635.51 USD
102110-770
Anti-SUB1 Rabbit Polyclonal Antibody
Antibodies
The SUB1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SUB1. This antibody reacts with human, mouse. The SUB1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 - 1:500

Type: Primary
Antigen: SUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR