Anti-STYK1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31419
102164-600EA 349 USD
102164-600
Anti-STYK1 Rabbit Polyclonal Antibody
The STYK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STYK1. This antibody reacts with human. The STYK1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STYK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR