Anti-Stxbp5l Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-91288
102119-046EA 564.74 USD
102119-046
Anti-Stxbp5l Rabbit Polyclonal Antibody
Antibodies
The Stxbp5l Antibody from Novus Biologicals is a rabbit polyclonal antibody to Stxbp5l. This antibody reacts with mouse. The Stxbp5l Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Stxbp5l
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR