Anti-STRAP Rabbit Polyclonal Antibody

NBP1-52949
102183-362EA 466.15 USD
102183-362
Anti-STRAP Rabbit Polyclonal Antibody
The STRAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to STRAP. This antibody reacts with human. The STRAP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: STRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR