Anti-Stomatin-like protein 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32227
102165-656EA 635.51 USD
102165-656
Anti-Stomatin-like protein 1 Rabbit Polyclonal Antibody
Antibodies
The Stomatin-like protein 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Stomatin-like protein 1. This antibody reacts with human. The Stomatin-like protein 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Stomatin-like protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR