Anti-STK39 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-30112
102162-970EA 635.51 USD
102162-970
Anti-STK39 Rabbit Polyclonal Antibody
Antibodies
The STK39 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK39. This antibody reacts with human. The STK39 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation (Negative).

Type: Primary
Antigen: STK39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: maps to a region between residue 425 and 450 of human STE20/SPS1-related proline-alanine-rich protein kinase using the numbering given in entry
AAC72238.1
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR