Anti-STK33 Rabbit Polyclonal Antibody

NBP1-32683
102166-242EA 493.85 USD
102166-242
Anti-STK33 Rabbit Polyclonal Antibody
The STK33 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK33. This antibody reacts with human. The STK33 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: STK33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR