Anti-STK32C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33507
102167-104EA 635.51 USD
102167-104
Anti-STK32C Rabbit Polyclonal Antibody
Antibodies
The STK32C Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK32C. This antibody reacts with human, mouse. The STK32C Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STK32C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR