Anti-STK25 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32670
102166-226EA 635.51 USD
102166-226
Anti-STK25 Rabbit Polyclonal Antibody
Antibodies
The STK25 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK25. This antibody reacts with human. The STK25 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STK25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR