|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | STIM2 |
Antigen Name | Stromal interaction molecule 2 |
Reactivity | Human, Mouse, Rat |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | KIAA1482, stromal interaction molecule 2, FLJ39527 |
Gene ID | 57620 |
Concentration | 1 mg/ml |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide from near the center of human STIM2. The immunogen is located within amino acids 140 - 190 of STIM2. |
Purification | Immunogen affinity purified |
Storage Buffer | PBS |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |