Anti-SF-1/NR5A1/Steroidogenic Factor 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-52823
102183-122EA 564.74 USD
102183-122
Anti-SF-1/NR5A1/Steroidogenic Factor 1 Rabbit Polyclonal Antibody
Antibodies
The SF-1 / NR5A1 / Steroidogenic Factor 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SF-1 / NR5A1 / Steroidogenic Factor 1. This antibody reacts with human. The SF-1 / NR5A1 / Steroidogenic Factor 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SF-1/NR5A1/Steroidogenic Factor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR