Anti-STEP Rabbit Polyclonal Antibody

NBP1-33388
102166-962EA 493.85 USD
102166-962
Anti-STEP Rabbit Polyclonal Antibody
The STEP Antibody from Novus Biologicals is a rabbit polyclonal antibody to STEP. This antibody reacts with human, mouse. The STEP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR