Anti-STAMP2/STEAP4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-80831
102118-384EA 686.46 USD
102118-384
Anti-STAMP2/STEAP4 Rabbit Polyclonal Antibody
Antibodies
The STAMP2 / STEAP4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STAMP2 / STEAP4. This antibody reacts with human, mouse, rat. The STAMP2 / STEAP4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: STAMP2/STEAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR