Anti-Staufen Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33202
102166-862EA 635.51 USD
102166-862
Anti-Staufen Rabbit Polyclonal Antibody
Antibodies
The Staufen Antibody from Novus Biologicals is a rabbit polyclonal antibody to Staufen. This antibody reacts with human, mouse. The Staufen Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: Staufen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR