Anti-Stathmin 1 Rabbit Polyclonal Antibody

NBP1-31748
102165-050EA 493.85 USD
102165-050
Anti-Stathmin 1 Rabbit Polyclonal Antibody
The Stathmin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Stathmin 1. This antibody reacts with human, mouse. The Stathmin 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Stathmin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR