Anti-STMN1 Rabbit Polyclonal Antibody

NB100-78407
102117-904EA 466.15 USD
102117-904
Anti-STMN1 Rabbit Polyclonal Antibody
Rabbit Polyclonal Stathmin 1 [p Ser16] Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR