Anti-STARD3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-599
102130-868EA 564.74 USD
102130-868
Anti-STARD3 Rabbit Polyclonal Antibody
Antibodies
The STARD3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STARD3. This antibody reacts with rat. The STARD3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: STARD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR